2-Isobutoxy-3-methylaniline 97%
Lieferant: Apollo Scientific
OR911545-1G
OR911545-5G
APOSOR911545-1GEA
151
EUR
APOSOR911545-1G
APOSOR911545-5G
2-Isobutoxy-3-methylaniline 97%
2-Isobutoxy-3-methylaniline
Formel:
C₁₁H₁₇NO Molecular Weight: 179.259 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD22205730 CAS-Nummer: 1375069-03-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...