2-Butoxy-3-methylaniline 97%
Lieferant: Apollo Scientific
OR911556-1G
OR911556-5G
APOSOR911556-1GEA
168
EUR
APOSOR911556-1G
APOSOR911556-5G
2-Butoxy-3-methylaniline 97%
2-Butoxy-3-methylaniline
Formel:
C₁₁H₁₇NO Molecular Weight: 179,259 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22205731 CAS-Nummer: 1375068-69-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...