4-Bromo-N-methyl piperidine 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR911812-1G
OR911812-5G
APOSOR911812-1GEA
78.3
EUR
APOSOR911812-1G
APOSOR911812-5G
4-Bromo-N-methyl piperidine 97%
4-Bromo-N-methyl piperidine
Formel:
C₆H₁₂BrN Lagertemperatur: Gefrierschrank |
MDL:
MFCD00800245 CAS-Nummer: 76444-51-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...