2-(2-Hydroxyphenyl)glycine 97%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-2-(2-hydroxyphenyl)acetic acid, H-DL-Phg(2-OH)-OH

OR911973-1G OR911973-5G
APOSOR911973-1GEA 393 EUR
APOSOR911973-1G APOSOR911973-5G
2-(2-Hydroxyphenyl)glycine 97%
2-(2-Hydroxyphenyl)glycine
Formel: C₈H₉NO₃
Molecular Weight: 167.162 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00014348
CAS-Nummer: 25178-38-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR