Pyrazolo[1,5-a]pyridine-3-carbaldehyde 95%

Lieferant: Apollo Scientific

OR912002-1G OR912002-250MG
APOSOR912002-1GEA 259 EUR
APOSOR912002-1G APOSOR912002-250MG
Pyrazolo[1,5-a]pyridine-3-carbaldehyde 95%
Pyrazolo[1,5-a]pyridine-3-carbaldehyde
Formel: C₈H₆N₂O
Molecular Weight: 146,146 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06739035
CAS-Nummer: 73957-66-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR