N-(4-Methoxybenzyl)cyclopropanamine 97%

Lieferant: Apollo Scientific

OR912024-5G
APOSOR912024-5GEA 149 EUR
APOSOR912024-5G
N-(4-Methoxybenzyl)cyclopropanamine 97%
N-(4-Methoxybenzyl)cyclopropanamine
Formel: C₁₁H₁₅NO
Molecular Weight: 177,243 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07310991
CAS-Nummer: 70894-71-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR