3,4,5,6-Tetrahydrophthalimide 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4,5,6,7-Tetrahydro-1H-isoindole-1,3(2H)-dione

OR912151-100MG OR912151-250MG
APOSOR912151-100MGEA 114 EUR
APOSOR912151-100MG APOSOR912151-250MG
3,4,5,6-Tetrahydrophthalimide 95%
3,4,5,6-Tetrahydrophthalimide
Formel: C₈H₉NO₂
Molecular Weight: 151,163 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01735122
CAS-Nummer: 4720-86-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR