(R)-1-N-CBZ-2-Methyl-piperazine 95%
Lieferant: Apollo Scientific
Synonyme:
(R)-1-Cbz-2-methylpiperazine
OR912199-1G
OR912199-250MG
OR912199-5G
APOSOR912199-1GEA
39.3
EUR
APOSOR912199-1G
APOSOR912199-250MG
APOSOR912199-5G
(R)-1-N-CBZ-2-Methyl-piperazine 95%
(R)-1-N-CBZ-2-Methyl-piperazine
Formel:
C₁₃H₁₈N₂O₂ Molecular Weight: 234.294 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115051 CAS-Nummer: 923565-99-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...