a-Amino-4-methoxybenzeneacetic acid 97%

Lieferant: Apollo Scientific

Synonyme: H-DL-Phg(4-OMe)-OH, 2-Amino-2-(4-methoxyphenyl)acetic acid

OR912250-1G OR912250-5G
APOSOR912250-1GEA 71.2 EUR
APOSOR912250-1G APOSOR912250-5G
a-Amino-4-methoxybenzeneacetic acid 97%
a-Amino-4-methoxybenzeneacetic acid
Formel: C₉H₁₁NO₃
Molecular Weight: 181.189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00049329
CAS-Nummer: 19789-59-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR