4-Methoxyindole-3-carboxaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: 4-Methoxy-1H-indole-3-carbaldehyde

OR912398-1G OR912398-5G
APOSOR912398-1GEA 119 EUR
APOSOR912398-1G APOSOR912398-5G
4-Methoxyindole-3-carboxaldehyde 95%
4-Methoxyindole-3-carboxaldehyde
Formel: C₁₀H₉NO₂
Molecular Weight: 175.184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00152191
CAS-Nummer: 90734-97-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR