trans-4-(Cbz-amino)cyclohexan-1-ol 97%
Lieferant: Apollo Scientific
Synonyme:
Benzyl (trans-4-hydroxycyclohexyl)carbamate
OR912410-25G
APOSOR912410-25GEA
127
EUR
APOSOR912410-25G
trans-4-(Cbz-amino)cyclohexan-1-ol 97%
trans-4-(Cbz-amino)cyclohexan-1-ol
Formel:
C₁₄H₁₉NO₃ Molecular Weight: 249,306 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00158431 CAS-Nummer: 27489-63-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...