N-Methyl-3-nitrobenzamide 98%
Lieferant: Apollo Scientific
OR912573-1G
OR912573-25G
OR912573-5G
APOSOR912573-1GEA
145
EUR
APOSOR912573-1G
APOSOR912573-25G
APOSOR912573-5G
N-Methyl-3-nitrobenzamide 98%
N-Methyl-3-nitrobenzamide
Formel:
C₈H₈N₂O₃ Molecular Weight: 180,161 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00714941 CAS-Nummer: 3400-26-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...