T-Butyl 3-chloropropanoate 95%

Lieferant: Apollo Scientific

APOSOR912679-1GEA 524 EUR
APOSOR912679-1G APOSOR912679-250MG
T-Butyl 3-chloropropanoate 95%
T-Butyl 3-chloropropanoate
Formel: C₇H₁₃ClO₂
Molecular Weight: 164,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01569942
CAS-Nummer: 55710-80-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR