2-Chloroisophthalonitrile 95%

Lieferant: Apollo Scientific

OR912695-1G OR912695-250MG
APOSOR912695-1GEA 244 EUR
APOSOR912695-1G APOSOR912695-250MG
2-Chloroisophthalonitrile 95%
2-Chloroisophthalonitrile
Formel: C₈H₃ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01657061
CAS-Nummer: 28442-78-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR