(S)-Methyl-4-chloro-3-hydroxybutyrate 96%
Lieferant: Apollo Scientific
Synonyme:
(S)-Methyl 4-chloro-3-hydroxybutanoate
OR912719-25G
OR912719-5G
APOSOR912719-25GEA
787
EUR
APOSOR912719-25G
APOSOR912719-5G
(S)-Methyl-4-chloro-3-hydroxybutyrate 96%
(S)-Methyl-4-chloro-3-hydroxybutyrate
Formel:
C₅H₉ClO₃ Molecular Weight: 152,576 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01862166 CAS-Nummer: 86728-93-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...