2-(2-Methoxyphenyl)glycine 95%

Lieferant: Apollo Scientific

Synonyme: H-DL-Phg(2-OMe)-OH, 2-Amino-2-(2-methoxyphenyl)acetic acid

OR912775-1G OR912775-5G
APOSOR912775-1GEA 115 EUR
APOSOR912775-1G APOSOR912775-5G
2-(2-Methoxyphenyl)glycine 95%
2-(2-Methoxyphenyl)glycine
Formel: C₉H₁₁NO₃
Molecular Weight: 181,189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02662408
CAS-Nummer: 271583-17-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR