4-(Propylamino)benzonitrile 98%

Lieferant: Apollo Scientific

OR912846-10G OR912846-1G OR912846-5G
APOSOR912846-10GEA 502 EUR
APOSOR912846-10G APOSOR912846-1G APOSOR912846-5G
4-(Propylamino)benzonitrile 98%
4-(Propylamino)benzonitrile
Formel: C₁₀H₁₂N₂
Molecular Weight: 160,216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11135495
CAS-Nummer: 4714-64-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR