4-(Isopropylamino)benzonitrile 98%

Lieferant: Apollo Scientific

OR912857-1G OR912857-5G
APOSOR912857-1GEA 219 EUR
APOSOR912857-1G APOSOR912857-5G
4-(Isopropylamino)benzonitrile 98%
4-(Isopropylamino)benzonitrile
Formel: C₁₀H₁₂N₂
Molecular Weight: 160.216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11145772
CAS-Nummer: 204078-26-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR