3,4-Diethyl-1H-pyrrole-2-carboxylic acid ethyl ester 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl 3,4-diethyl-1H-pyrrole-2-carboxylate

OR912894-1G OR912894-5G
APOSOR912894-1GEA 223 EUR
APOSOR912894-1G APOSOR912894-5G
3,4-Diethyl-1H-pyrrole-2-carboxylic acid ethyl ester 95%
3,4-Diethyl-1H-pyrrole-2-carboxylic acid ethyl ester
Formel: C₁₁H₁₇NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03453113
CAS-Nummer: 97336-41-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR