1-(3-amino-2-methylphenyl)pyrrolidin-2-one 98%

Lieferant: Apollo Scientific

OR912925-1G OR912925-5G
APOSOR912925-1GEA 270 EUR
APOSOR912925-1G APOSOR912925-5G
1-(3-amino-2-methylphenyl)pyrrolidin-2-one 98%
1-(3-amino-2-methylphenyl)pyrrolidin-2-one
Formel: C₁₁H₁₄N₂O
Molecular Weight: 190,242 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09044451
CAS-Nummer: 69131-43-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR