6-Methoxy-1H-indazole-3-carboxylic acid 97+%
Lieferant: Apollo Scientific
|
Achtung
|
OR912929-1G
OR912929-250MG
APOSOR912929-1GEA
223
EUR
APOSOR912929-1G
APOSOR912929-250MG
6-Methoxy-1H-indazole-3-carboxylic acid 97+%
6-Methoxy-1H-indazole-3-carboxylic acid
Formel:
C₉H₈N₂O₃ Molecular Weight: 192.171 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD03840714 CAS-Nummer: 518990-36-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...