7-Methoxy-1H-indazole-3-carboxylic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR913077-1G
OR913077-250MG
APOSOR913077-1GEA
493
EUR
APOSOR913077-1G
APOSOR913077-250MG
7-Methoxy-1H-indazole-3-carboxylic acid 95%
7-Methoxy-1H-indazole-3-carboxylic acid
Formel:
C₉H₈N₂O₃ Molecular Weight: 192,171 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD05663994 CAS-Nummer: 133841-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...