3-Methoxyphenylglyoxal hydrate 96%

Lieferant: Apollo Scientific

Synonyme: (3-Methoxyphenyl)(oxo)acetaldehyde

OR913084-1G OR913084-5G
APOSOR913084-1GEA 116 EUR
APOSOR913084-1G APOSOR913084-5G
3-Methoxyphenylglyoxal hydrate 96%
3-Methoxyphenylglyoxal hydrate
Formel: C₉H₈O₃·xH₂O
Molecular Weight: 164,16 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD05664093
CAS-Nummer: 1201806-19-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR