(3,5-Dimethoxybenzyl)methylamine 95%
Lieferant: Apollo Scientific
Synonyme:
1-(3,5-Dimethoxyphenyl)-N-methylmethanamine
OR913105-1G
OR913105-5G
APOSOR913105-1GEA
60.9
EUR
APOSOR913105-1G
APOSOR913105-5G
(3,5-Dimethoxybenzyl)methylamine 95%
(3,5-Dimethoxybenzyl)methylamine
Formel:
C₁₀H₁₅NO₂ Molecular Weight: 181,232 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05864733 CAS-Nummer: 77775-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...