5-Chloro-1H-indazole-3-carbonitrile 95%

Lieferant: Apollo Scientific
Achtung

OR913233-100MG OR913233-1G OR913233-250MG
APOSOR913233-100MGEA 31.9 EUR
APOSOR913233-100MG APOSOR913233-1G APOSOR913233-250MG
5-Chloro-1H-indazole-3-carbonitrile 95%
5-Chloro-1H-indazole-3-carbonitrile
Formel: C₈H₄ClN₃
Molecular Weight: 177,59 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07371575
CAS-Nummer: 29646-35-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR