7-Methylindoline 95%

Lieferant: Apollo Scientific

OR913234-1G OR913234-250MG OR913234-5G
APOSOR913234-1GEA 306 EUR
APOSOR913234-1G APOSOR913234-250MG APOSOR913234-5G
7-Methylindoline 95%
7-Methylindoline
Formel: C₉H₁₁N
Molecular Weight: 133,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07371631
CAS-Nummer: 65673-86-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR