1-(5-Chloro-1H-indol-3-yl)ethanone 95%

Lieferant: Apollo Scientific

OR913238-1G OR913238-250MG
APOSOR913238-1GEA 122 EUR
APOSOR913238-1G APOSOR913238-250MG
1-(5-Chloro-1H-indol-3-yl)ethanone 95%
1-(5-Chloro-1H-indol-3-yl)ethanone
Formel: C₁₀H₈ClNO
Molecular Weight: 193,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07437837
CAS-Nummer: 51843-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR