3-Methyl-4-piperidone 95%
Lieferant: Apollo Scientific
Synonyme:
3-Methylpiperidin-4-one
OR913284-1G
OR913284-250MG
APOSOR913284-1GEA
108
EUR
APOSOR913284-1G
APOSOR913284-250MG
3-Methyl-4-piperidone 95%
3-Methyl-4-piperidone
Formel:
C₆H₁₁NO Molecular Weight: 113,158 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07787596 CAS-Nummer: 5773-58-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...