Benzo[b]thiophene-3-carbonitrile 95%

Lieferant: Apollo Scientific

OR913339-1G OR913339-5G
APOSOR913339-1GEA 55.1 EUR
APOSOR913339-1G APOSOR913339-5G
Benzo[b]thiophene-3-carbonitrile 95%
Benzo[b]thiophene-3-carbonitrile
Formel: C₉H₅NS
Molecular Weight: 159.208 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08276872
CAS-Nummer: 24434-84-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR