5,6-Dihydro-4h-pyran-2-carbonitrile 95%
Lieferant: Apollo Scientific
OR913345-1G
OR913345-250MG
APOSOR913345-1GEA
194
EUR
APOSOR913345-1G
APOSOR913345-250MG
5,6-Dihydro-4h-pyran-2-carbonitrile 95%
5,6-Dihydro-4h-pyran-2-carbonitrile
Formel:
C₆H₇NO Molecular Weight: 109,126 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08437233 CAS-Nummer: 31518-13-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...