5-Bromo-N1-methylbenzene-1,2-diamine 96%

Lieferant: Apollo Scientific
Achtung

OR913515-1G OR913515-250MG OR913515-5G
APOSOR913515-1GEA 49.3 EUR
APOSOR913515-1G APOSOR913515-250MG APOSOR913515-5G
5-Bromo-N1-methylbenzene-1,2-diamine 96%
5-Bromo-N1-methylbenzene-1,2-diamine
Formel: C₇H₉BrN₂
Molecular Weight: 201.064 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11977373
CAS-Nummer: 337915-79-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR