(4-Chloro-2-formyl-phenoxy)-acetonitrile 95%
Lieferant: Apollo Scientific
OR913532-1G
OR913532-250MG
APOSOR913532-1GEA
647
EUR
APOSOR913532-1G
APOSOR913532-250MG
(4-Chloro-2-formyl-phenoxy)-acetonitrile 95%
(4-Chloro-2-formyl-phenoxy)-acetonitrile
Formel:
C₉H₆ClNO₂ Molecular Weight: 195,602 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07431460 CAS-Nummer: 125418-96-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...