2-Chloro-1,3-dimethyl-5-nitrobenzene 97%

Lieferant: Apollo Scientific

OR913580-1G OR913580-25G OR913580-5G
APOSOR913580-1GEA 132 EUR
APOSOR913580-1G APOSOR913580-25G APOSOR913580-5G
2-Chloro-1,3-dimethyl-5-nitrobenzene 97%
2-Chloro-1,3-dimethyl-5-nitrobenzene
Formel: C₈H₈ClNO₂
Molecular Weight: 185,608 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02178895
CAS-Nummer: 38560-96-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR