(R)-1-N-Benzyl-prolinol 95%
Lieferant: Apollo Scientific
OR913624-1G
OR913624-5G
APOSOR913624-1GEA
174
EUR
APOSOR913624-1G
APOSOR913624-5G
(R)-1-N-Benzyl-prolinol 95%
(R)-1-N-Benzyl-prolinol
Formel:
C₁₂H₁₇NO Molecular Weight: 191,269 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03790915 CAS-Nummer: 182076-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...