3,5-Dibromo-4-methoxyaniline

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-dibromo-4-methoxyaniline

OR913648-1G OR913648-5G
APOSOR913648-1GEA 299 EUR
APOSOR913648-1G APOSOR913648-5G
3,5-Dibromo-4-methoxyaniline
3,5-Dibromo-4-methoxyaniline
Formel: C₇H₇Br₂NO
Molecular Weight: 280,945 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11647741
CAS-Nummer: 41727-70-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR