(R)-3-Amino-3-(4-methylphenyl)propionic acid 97%

Lieferant: Apollo Scientific

Synonyme: (R)-3-(p-Methylphenyl)-beta-alanine

OR913672-1G OR913672-5G
APOSOR913672-1GEA 106 EUR
APOSOR913672-1G APOSOR913672-5G
(R)-3-Amino-3-(4-methylphenyl)propionic acid 97%
(R)-3-Amino-3-(4-methylphenyl)propionic acid
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03840389
CAS-Nummer: 479064-87-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR