(R)-3-Amino-3-(3-bromophenyl)propionic acid 97%

Lieferant: Apollo Scientific

Synonyme: (R)-3-Amino-3-(3-bromophenyl)propanoic acid

OR913675-1G
APOSOR913675-1GEA 75.4 EUR
APOSOR913675-1G
(R)-3-Amino-3-(3-bromophenyl)propionic acid 97%
(R)-3-Amino-3-(3-bromophenyl)propionic acid
Formel: C₉H₁₀BrNO₂
Molecular Weight: 244,085 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04113653
CAS-Nummer: 788153-27-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR