(R)-3-Amino-3-(3-methylphenyl)propionic acid 97%

Lieferant: Apollo Scientific

Synonyme: (R)-3-Amino-3-(m-tolyl)propanoic acid

OR913676-1G OR913676-5G
APOSOR913676-1GEA 104 EUR
APOSOR913676-1G APOSOR913676-5G
(R)-3-Amino-3-(3-methylphenyl)propionic acid 97%
(R)-3-Amino-3-(3-methylphenyl)propionic acid
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179.216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04113659
CAS-Nummer: 748128-33-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR