(R)-3-Amino-3-(4-methoxyphenyl)propionic acid 97%
Lieferant: Apollo Scientific
Synonyme:
(R)-3-Amino-3-(4-methoxyphenyl)propanoic acid
OR913679-1G
APOSOR913679-1GEA
150
EUR
APOSOR913679-1G
(R)-3-Amino-3-(4-methoxyphenyl)propionic acid 97%
(R)-3-Amino-3-(4-methoxyphenyl)propionic acid
Formel:
C₁₀H₁₃NO₃ Molecular Weight: 195,215 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01318245 CAS-Nummer: 131690-57-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...