Methyl (3R)-3-aminobutanoate hydrochloride 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (R)-3-Amino-butyric acid methyl ester hydrochloride, (R)-Methyl 3-aminobutanoate hydrochloride

OR913686-1G OR913686-25G OR913686-5G
APOSOR913686-1GEA 118 EUR
APOSOR913686-1G APOSOR913686-25G APOSOR913686-5G
Methyl (3R)-3-aminobutanoate hydrochloride 98%
Methyl (3R)-3-aminobutanoate hydrochloride
Formel: C₅H₁₁NO₂·ClH
Molecular Weight: 153.607 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12068616
CAS-Nummer: 139243-54-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR