2,3-Dihydro-5-[(methylamino)methyl]thieno[3,4-b][1,4]dioxine 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
N-(2,3-Dihydrothieno[3,4-b][1,4]dioxin-5-ylmethyl)-N-methylamine
OR9137-1G
OR9137-250MG
APOSOR9137-1GEA
197
EUR
APOSOR9137-1G
APOSOR9137-250MG
2,3-Dihydro-5-[(methylamino)methyl]thieno[3,4-b][1,4]dioxine 95%
2,3-Dihydro-5-[(methylamino)methyl]thieno[3,4-b][1,4]dioxine
Formel:
C₈H₁₁NO₂S Molecular Weight: 185,243 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08271957 CAS-Nummer: 859851-03-9 UN: 2922 ADR: 8+6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...