(R)-4-Cyanothiazolidine hydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: (R)-Thiazolidine-4-carbonitrile hydrochloride

OR913722-1G OR913722-250MG
APOSOR913722-1GEA 574 EUR
APOSOR913722-1G APOSOR913722-250MG
(R)-4-Cyanothiazolidine hydrochloride 95%
(R)-4-Cyanothiazolidine hydrochloride
Formel: C₄H₆N₂S·ClH
Molecular Weight: 150.63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02683061
CAS-Nummer: 391248-17-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR