(S)-1,4-Benzodioxane-2-carboxylic acid 95%

Lieferant: Apollo Scientific

OR913812-1G OR913812-250MG
APOSOR913812-1GEA 89.9 EUR
APOSOR913812-1G APOSOR913812-250MG
(S)-1,4-Benzodioxane-2-carboxylic acid 95%
(S)-1,4-Benzodioxane-2-carboxylic acid
Formel: C₉H₈O₄
Molecular Weight: 180,157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00239408
CAS-Nummer: 70918-54-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR