(S)-1-N-Benzyl-2-cyano-pyrrolidine 95%

Lieferant: Apollo Scientific

OR913834-1G OR913834-250MG
APOSOR913834-1GEA 415 EUR
APOSOR913834-1G APOSOR913834-250MG
(S)-1-N-Benzyl-2-cyano-pyrrolidine 95%
(S)-1-N-Benzyl-2-cyano-pyrrolidine
Formel: C₁₂H₁₄N₂
Molecular Weight: 186.253 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05864626
CAS-Nummer: 928056-25-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR