(S)-3-Amino-butyric acid methyl ester hydrochloride 96%

Lieferant: Apollo Scientific

Synonyme: (S)-Methyl 3-aminobutanoate hydrochloride

OR913942-1G OR913942-250MG
APOSOR913942-1GEA 95.7 EUR
APOSOR913942-1G APOSOR913942-250MG
(S)-3-Amino-butyric acid methyl ester hydrochloride 96%
(S)-3-Amino-butyric acid methyl ester hydrochloride
Formel: C₅H₁₁NO₂·ClH
Molecular Weight: 153,607 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12068617
CAS-Nummer: 139243-55-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR