(S)-4-Benzyl-1,3-thiazolidine-2-one 97%

Lieferant: Apollo Scientific

OR913966-1G OR913966-250MG OR913966-5G
APOSOR913966-1GEA 255 EUR
APOSOR913966-1G APOSOR913966-250MG APOSOR913966-5G
(S)-4-Benzyl-1,3-thiazolidine-2-one 97%
(S)-4-Benzyl-1,3-thiazolidine-2-one
Formel: C₁₀H₁₁NOS
Molecular Weight: 193,265 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08436194
CAS-Nummer: 219821-18-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR