(S)-4-Boc-2-(aminomethyl)morpholine 97%
Lieferant: Apollo Scientific
Synonyme:
(S)-tert-Butyl 2-(aminomethyl)morpholine-4-carboxylate
OR914006-1G
OR914006-250MG
APOSOR914006-1GEA
97.3
EUR
APOSOR914006-1G
APOSOR914006-250MG
(S)-4-Boc-2-(aminomethyl)morpholine 97%
(S)-4-Boc-2-(aminomethyl)morpholine
Formel:
C₁₀H₂₀N₂O₃ Molecular Weight: 216,277 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11707017 CAS-Nummer: 879403-42-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...