[4-(Propan-2-yloxy)phenyl]methanol 97%
Lieferant: Apollo Scientific
OR914059-1G
OR914059-5G
APOSOR914059-1GEA
248
EUR
APOSOR914059-1G
APOSOR914059-5G
[4-(Propan-2-yloxy)phenyl]methanol 97%
[4-(Propan-2-yloxy)phenyl]methanol
Formel:
C₁₀H₁₄O₂ Molecular Weight: 166,217 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06202646 CAS-Nummer: 82657-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...