1-(2-Hydroxyethyl)-4-isopropylpiperazine 97%
Lieferant: Apollo Scientific
OR914084-1G
OR914084-5G
APOSOR914084-1GEA
244
EUR
APOSOR914084-1G
APOSOR914084-5G
1-(2-Hydroxyethyl)-4-isopropylpiperazine 97%
1-(2-Hydroxyethyl)-4-isopropylpiperazine
Formel:
C₉H₂₀N₂O Molecular Weight: 172,268 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06740831 CAS-Nummer: 103069-50-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...