Methyl 1-(4-bromophenyl)cyclobutane-1-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 1-(4-bromophenyl)cyclobutanecarboxylate

OR914109-1G OR914109-250MG OR914109-5G
APOSOR914109-1GEA 78.3 EUR
APOSOR914109-1G APOSOR914109-250MG APOSOR914109-5G
Methyl 1-(4-bromophenyl)cyclobutane-1-carboxylate
Methyl 1-(4-bromophenyl)cyclobutane-1-carboxylate
Formel: C₁₂H₁₃BrO₂
Molecular Weight: 269,134 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18711569
CAS-Nummer: 1236357-65-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR